Alfa Aesar™ Bis(tetramethylcyclopentadienyl)manganese(II), 98+%

Manufacturer: Alfa Aesar™ H2702706

Quantity 5g

 View more versions of this product

Catalog No. AAH2702706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 101932-75-6
    Molecular Formula C18H26Mn
    Formula Weight 297.35
    MDL Number MFCD01862454
    Chemical Name or Material Bis(tetramethylcyclopentadienyl)manganese(II)

    Specifications

    Formula Weight 297.35
    Molecular Formula C18H26Mn
    MDL Number MFCD01862454
    CAS 101932-75-6
    Melting Point 98° to 112°C
    Quantity 5g
    Recommended Storage 2° to 8°C
    Chemical Name or Material Bis(tetramethylcyclopentadienyl)manganese(II)
    Assay ≥98%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit