Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis[tri(o-tolyl)phosphine]palladium(II) chloride, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
40691-33-6
C42H42Cl2P2Pd
786.06
MFCD00274659
KFQYTZDYGOBBOE-UHFFFAOYSA-L
palladium,dichlorobis tris 2-methylphenyl phosphine
90656943
palladium;tris(2-methylphenyl)phosphane;dichloride
CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C.CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C.[Cl-].[Cl-].[Pd]
40691-33-6
C42H42Cl2P2Pd
786.06
MFCD00274659
KFQYTZDYGOBBOE-UHFFFAOYSA-L
palladium,dichlorobis tris 2-methylphenyl phosphine
90656943
palladium;tris(2-methylphenyl)phosphane;dichloride
CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C.CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C.[Cl-].[Cl-].[Pd]
Melting Point 280°C
Quantity 5g
Color Yellow to Orange
Packaging Glass bottle
Infrared Spectrum Authentic
Elemental Analysis 94% min. (ex Carbon)
Assay Percent Range 95%
Solubility Information Solubility in water: insoluble
Formula Weight 786.06
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material Bis[tri(o-tolyl)phosphine]palladium(II) chloride
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning