Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(tributylstannyl)acetylene, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
994-71-8
C26H54Sn2
604.13
MFCD00010239
CAUONNQGFXOEMY-UHFFFAOYSA-N
bis tributylstannyl acetylene,1,2-bis tributylstannyl ethyne,bis tri-n-butylstannyl acetylene,bis tri-n-butylstannyl ethyne,tributyl 2-tributylstannylethynyl stannane,tributyl tributylstannyl ethynyl stannane #,tributyl 2-tributylstannyl ethynyl stannane
305981
tributyl(2-tributylstannylethynyl)stannane
CCCC[Sn](CCCC)(CCCC)C#C[Sn](CCCC)(CCCC)CCCC
994-71-8
C26H54Sn2
604.13
MFCD00010239
CAUONNQGFXOEMY-UHFFFAOYSA-N
bis tributylstannyl acetylene,1,2-bis tributylstannyl ethyne,bis tri-n-butylstannyl acetylene,bis tri-n-butylstannyl ethyne,tributyl 2-tributylstannylethynyl stannane,tributyl tributylstannyl ethynyl stannane #,tributyl 2-tributylstannyl ethynyl stannane
305981
tributyl(2-tributylstannylethynyl)stannane
CCCC[Sn](CCCC)(CCCC)C#C[Sn](CCCC)(CCCC)CCCC
Specific Gravity 1.147
Color Colorless
Flash Point >110°C
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4920 to 1.4940
Density 1.1470g/mL
Assay Percent Range 96% min. (GC)
Linear Formula [CH3(CH2)3]3SnC≡CSn[(CH2)3CH3]3
Formula Weight 604.13
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Bis(tributylstannyl)acetylene
Show More Show Less

GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with

GHS P Statement
Take off contaminated clothing and wash before reuse.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash wi

GHS Signal Word: Danger