Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(tricarbonylcyclopentadienylmolybdenum)

Manufacturer:  Alfa Aesar™ 4812103

Quantity 1g

 View more versions of this product

Catalog No. AA4812103

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

12091-64-4
490.13
UN3466
Bis (tricarbonylcyclopentadienylmolybdenum)
C16H10Mo2O6
MFCD00009947
Cyclopentadienylmolybdenum tricarbonyl dimer; Tricarbonylcyclopentadienylmolybdenum dimer

Specifications

12091-64-4
C16H10Mo2O6
MFCD00009947
Cyclopentadienylmolybdenum tricarbonyl dimer; Tricarbonylcyclopentadienylmolybdenum dimer
1g
Crystalline
490.13
UN3466
180°C (dec.)
Bis (tricarbonylcyclopentadienylmolybdenum)

Safety and Handling

GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit