Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(triethylphosphine)palladium(II) chloride, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
28425-04-9
C12H30Cl2P2Pd
413.64
MFCD00191831
ULYNIEUXPCUIEL-UHFFFAOYSA-L
dichlorobis triethylphosphine palladium ii,bis triethylphosphine palladium dichloride,trans-dichlorobis triethylphosphine palladium ii,bis triethylphosphine palladium ii chloride,pdcl2 pet3 2,palladium,dichlorobis triethylphosphine,dichloropalladium-triethylphosphane 1/2,bis triethylphosphine dichloridopalladium ii
11144124
dichloropalladium;triethylphosphane
CCP(CC)CC.CCP(CC)CC.Cl[Pd]Cl
28425-04-9
C12H30Cl2P2Pd
413.64
MFCD00191831
ULYNIEUXPCUIEL-UHFFFAOYSA-L
dichlorobis triethylphosphine palladium ii,bis triethylphosphine palladium dichloride,trans-dichlorobis triethylphosphine palladium ii,bis triethylphosphine palladium ii chloride,pdcl2 pet3 2,palladium,dichlorobis triethylphosphine,dichloropalladium-triethylphosphane 1/2,bis triethylphosphine dichloridopalladium ii
11144124
dichloropalladium;triethylphosphane
CCP(CC)CC.CCP(CC)CC.Cl[Pd]Cl
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
Formula Weight 413.64
Physical Form Powder
Chemical Name or Material Bis(triethylphosphine)palladium(II) chloride

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning