Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(trifluoromethanesulfonyl)imide, 99%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
82113-65-3
C2HF6NO4S2
281.16
MFCD00214154
ZXMGHDIOOHOAAE-UHFFFAOYSA-N
1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide,trifluoromethanesulfonimide,unii-c7r849vm9l,bis trifluoromethanesulfonyl imide,bistrifluoromethanesulfonimide,1,1,1-trifluoro-n-trifluoromethanesulfonylmethanesulfonamide,1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide,bis trifluoromethane sulfonimide,n,n-bis trifluoromethanesulfonyl imide,n,n-bis trifluoromethylsulphonyl amine
157857
1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
82113-65-3
C2HF6NO4S2
281.16
MFCD00214154
ZXMGHDIOOHOAAE-UHFFFAOYSA-N
1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide,trifluoromethanesulfonimide,unii-c7r849vm9l,bis trifluoromethanesulfonyl imide,bistrifluoromethanesulfonimide,1,1,1-trifluoro-n-trifluoromethanesulfonylmethanesulfonamide,1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide,bis trifluoromethane sulfonimide,n,n-bis trifluoromethanesulfonyl imide,n,n-bis trifluoromethylsulphonyl amine
157857
1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
Coupling reagent
Melting Point 52°C to 56°C
Boiling Point 90°C to 91°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Brown to Colorless
Quantity 25g
Assay Percent Range 99%
Solubility Information Solubility in water: reacts
Formula Weight 281.16
Physical Form Crystals
Percent Purity 99%
Chemical Name or Material Bis(trifluoromethanesulfonyl)imide
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger