Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(trimethylsilyl)acetylene, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
14630-40-1
C8H18Si2
170.41
MFCD00008276
ZDWYFWIBTZJGOR-UHFFFAOYSA-N
bis trimethylsilyl acetylene, silane, 1,2-ethynediylbis trimethyl, 1,2-bis trimethylsilyl ethyne, btmsa, 1,2-bis-trimethylsilanyl-ethyne, ethyne-1,2-diylbis trimethylsilane, 2,5-disilahex-3-yne, 2,2,5,5-tetramethyl, trimethyl 2-trimethylsilyl ethynyl silane, silane, 1,1'-1,2-ethynediyl bis 1,1,1-trimethyl
84564
trimethyl(2-trimethylsilylethynyl)silane
C[Si](C)(C)C#C[Si](C)(C)C
14630-40-1
C8H18Si2
170.41
MFCD00008276
ZDWYFWIBTZJGOR-UHFFFAOYSA-N
bis trimethylsilyl acetylene, silane, 1,2-ethynediylbis trimethyl, 1,2-bis trimethylsilyl ethyne, btmsa, 1,2-bis-trimethylsilanyl-ethyne, ethyne-1,2-diylbis trimethylsilane, 2,5-disilahex-3-yne, 2,2,5,5-tetramethyl, trimethyl 2-trimethylsilyl ethynyl silane, silane, 1,1'-1,2-ethynediyl bis 1,1,1-trimethyl
84564
trimethyl(2-trimethylsilylethynyl)silane
C[Si](C)(C)C#C[Si](C)(C)C
Specific Gravity 0.75
Packaging Glass bottle
Refractive Index 1.4260 to 1.4280
Color Colorless
Density 0.7500g/mL
Infrared Spectrum Authentic
Flash Point 2°C
Melting Point 21.0°C to 24.0°C
Boiling Point 136.0°C to 137.0°C
Quantity 50g
Assay Percent Range 98.5% min. (GC)
Linear Formula (CH3)3SiC=CSi(CH3)3
Fieser 05,44; 13,97
Solubility Information Solubility in water: may decompose
Formula Weight 170.41
Physical Form Liquid After Melting
Percent Purity 99%
Chemical Name or Material Bis(trimethylsilyl)acetylene, 99%
Show More Show Less

GHS H Statement
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Danger

EINECSNumber : 238-671-9

TSCA : TSCA