Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bis(trimethylsilyl)carbodiimide, 98%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
Description
Coupling reagent
Specifications
Density | 0.8210g/mL |
Refractive Index | 1.4330 to 1.435 |
Melting Point | -23°C |
Boiling Point | 164°C |
Flash Point | 42°C |
Specific Gravity | 0.821 |
Color | Beige to Yellow or White |
Packaging | Glass bottle |
Quantity | 1g |
Infrared Spectrum | Authentic |
Show More |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 213-673-2