Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(trimethylsilyl)carbodiimide, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1000-70-0
C7H18N2Si2
186.4
MFCD00051538
KSVMTHKYDGMXFJ-UHFFFAOYSA-N
bis trimethylsilyl carbodiimide,carbodiimide, bis trimethylsilyl,n,n'-methanediylidenebis 1,1,1-trimethylsilanamine,silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl,1,3-bis trimethylsilyl carbodiimide,n,n'-bis trimethylsilyl carbodiimide,n,n'-methane tetraylbis 1,1,1-trimethylsilanamine,carbodiimide, bis trimethylsilyl-7ci,8ci,n,n'-methanetetraylbis 1,1,1-trimethylsilylamine
70473
N,N'-bis(trimethylsilyl)methanediimine
C[Si](C)(C)N=C=N[Si](C)(C)C
1000-70-0
C7H18N2Si2
186.4
MFCD00051538
KSVMTHKYDGMXFJ-UHFFFAOYSA-N
bis trimethylsilyl carbodiimide,carbodiimide, bis trimethylsilyl,n,n'-methanediylidenebis 1,1,1-trimethylsilanamine,silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl,1,3-bis trimethylsilyl carbodiimide,n,n'-bis trimethylsilyl carbodiimide,n,n'-methane tetraylbis 1,1,1-trimethylsilanamine,carbodiimide, bis trimethylsilyl-7ci,8ci,n,n'-methanetetraylbis 1,1,1-trimethylsilylamine
70473
N,N'-bis(trimethylsilyl)methanediimine
C[Si](C)(C)N=C=N[Si](C)(C)C
Coupling reagent
Density 0.8210g/mL
Refractive Index 1.4330 to 1.435
Melting Point -23°C
Flash Point 42°C
Boiling Point 164°C
Specific Gravity 0.821
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Color Beige to Yellow or White
Assay Percent Range 98%
Solubility Information Solubility in water: insoluble
Formula Weight 186.4
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity ≥97.5% (GC)
Chemical Name or Material Bis(trimethylsilyl)carbodiimide
Show More Show Less

GHS P Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning