Alfa Aesar™ Bis(trimethylsilyl) sulfate, 97%

Manufacturer: Alfa Aesar™ B2360822

Quantity 100g

 View more versions of this product

Catalog No. AAB2360822

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 18306-29-1
    Molecular Formula C12H18O4SSi2
    Linear Formula [(CH3)3SiO]2SO2
    Formula Weight 242.44
    MDL Number MFCD00008268
    UN Number UN2925
    Chemical Name or Material Bis(trimethylsilyl) sulfate

    Specifications

    Formula Weight 242.44
    Linear Formula [(CH3)3SiO]2SO2
    MDL Number MFCD00008268
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    Molecular Formula C12H18O4SSi2
    CAS 18306-29-1
    UN Number UN2925
    Boiling Point 87° to 90°C (4mmHg)
    Melting Point 44° to 47°C
    Quantity 100g
    Flash Point −2°C (27°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H228-H314-H318
    Flammable solid.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material Bis(trimethylsilyl) sulfate
    Assay 97%

    GHS H Statement
    H228-H314-H318
    Flammable solid.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit