Alfa Aesar™ Bis(trimethylsilyl)sulfide, tech.

Manufacturer: Alfa Aesar™ 4481806

Quantity 5g

 View more versions of this product

Catalog No. AA4481806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3385-94-2
    Molecular Formula C6H18SSi2
    Linear Formula [(CH3)3Si]2S
    Formula Weight 178.44
    MDL Number MFCD00014851
    UN Number UN1993
    Beilstein 1740046
    Synonym Hexamethyldisilathiane
    Chemical Name or Material Bis (trimethylsilyl)sulfide
    Grade Technical Grade

    Specifications

    Formula Weight 178.44
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 1740046
    Synonym Hexamethyldisilathiane
    Boiling Point 162° to 166°C
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Physical Form Liquid
    Health Hazard 1 Warning
    Odor Pungent
    Sensitivity Air Sensitive
    Density 0.851g/mL
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Molecular Formula C6H18SSi2
    Linear Formula [(CH3)3Si]2S
    MDL Number MFCD00014851
    CAS 3385-94-2
    Color Colorless
    Refractive Index 1.4598
    UN Number UN1993
    Flash Point 26°C (79°F)
    Chemical Name or Material Bis (trimethylsilyl)sulfide
    Grade Technical Grade

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit