Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α,α-Bis(trimethylsilyl)-tert-butylketimine, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass ampule with screw-cap
147227-26-7
C12H29NSi2
243.541
MFCD01632578
KNFKVWSSVOLVHY-UHFFFAOYSA-N
3,3-dimethyl-1,1-bis trimethylsilyl butan-2-imine
90470112
3,3-dimethyl-1,1-bis(trimethylsilyl)butan-2-imine
CC(C)(C)C(=N)C([Si](C)(C)C)[Si](C)(C)C
147227-26-7
C12H29NSi2
243.541
MFCD01632578
KNFKVWSSVOLVHY-UHFFFAOYSA-N
3,3-dimethyl-1,1-bis trimethylsilyl butan-2-imine
90470112
3,3-dimethyl-1,1-bis(trimethylsilyl)butan-2-imine
CC(C)(C)C(=N)C([Si](C)(C)C)[Si](C)(C)C
Quantity 500mg
Packaging Glass ampule with screw-cap
Color Yellow
Infrared Spectrum Authentic
Refractive Index 1.4505 to 1.4525 (20°C, 589nm)
NMR Complies
Formula Weight 257.57
Physical Form Liquid
Chemical Name or Material α, α-Bis(trimethylsilyl)-tert-butylketimine