Alfa Aesar™ Bis(triphenyltin) oxide, 98%

Manufacturer: Alfa Aesar™ B2360906

Quantity 5g

 View more versions of this product

Catalog No. AAB2360906

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 1262-21-1
    Molecular Formula [(C6H5)3Sn]2O
    Formula Weight 716.06
    MDL Number MFCD00003005
    UN Number UN3146
    Beilstein 4166537
    Synonym Hexaphenyl distannoxane
    Chemical Name or Material Bis(triphenyltin) oxide

    Specifications

    Formula Weight 716.06
    MDL Number MFCD00003005
    Quantity 5g
    UN Number UN3146
    Beilstein 4166537
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 119° to 123°C
    Molecular Formula [(C6H5)3Sn]2O
    CAS 1262-21-1
    Synonym Hexaphenyl distannoxane
    Health Hazard 2 GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
    Chemical Name or Material Bis(triphenyltin) oxide
    Assay 98%

    GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit