Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ BIS-TRIS, 0.2M buffer soln., pH 7.4, low endotoxin

Click to view available options
Quantity:
100mL
250mL
6976-37-0
C8H19NO5
209.242
MFCD00002853
OWMVSZAMULFTJU-UHFFFAOYSA-N
81462
CHEBI:41250
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C(CO)N(CCO)C(CO)(CO)CO
6976-37-0
C8H19NO5
209.242
MFCD00002853
OWMVSZAMULFTJU-UHFFFAOYSA-N
81462
CHEBI:41250
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C(CO)N(CCO)C(CO)(CO)CO
Concentration 0.2M buffer soln.
Physical Form Liquid
pH 7.4
Color Colorless
Quantity 250mL
Chemical Name or Material BIS-TRIS, low endotoxin

TSCA : TSCA

storageNote1 : 2° to 8°C