Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Bis-Tris, ≥98% (Dry Basis), Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
100g
500g
1kg
Packaging:
Plastic bottle
6976-37-0
C8H19NO5
209.242
MFCD00002853
OWMVSZAMULFTJU-UHFFFAOYSA-N
bis[2 to Hydroxyethyl]imino to tris[hydroxymethyl]methane
81462
CHEBI:41250
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C(CO)N(CCO)C(CO)(CO)CO
6976-37-0
C8H19NO5
209.242
MFCD00002853
OWMVSZAMULFTJU-UHFFFAOYSA-N
bis[2 to Hydroxyethyl]imino to tris[hydroxymethyl]methane
81462
CHEBI:41250
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C(CO)N(CCO)C(CO)(CO)CO
Linear Formula (HOCH2CH2)2NC(CH2OH)3
Formula Weight 209.24
Physical Form Crystalline Powder
Percent Purity ≥98%
Packaging Plastic bottle
Melting Point 102°C to 105°C
Quantity 1kg
Chemical Name or Material bis[2-Hydroxyethyl]imino-tris[hydroxymethyl]methane
Assay Dry basis