Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis-Tris, cGMP Manufactured, 99%, Spectrum™

Click to view available options
Quantity:
1 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Drum
Poly Pail
6976-37-0
C8H19NO5
209.24
OWMVSZAMULFTJU-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
OCCN(CCO)C(CO)(CO)CO
6976-37-0
C8H19NO5
209.24
OWMVSZAMULFTJU-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
OCCN(CCO)C(CO)(CO)CO

B4092, 6976-37-0, C8H19NO5

Spectrum™ Bis-Tris, cGMP Manufactured - Spectrum cGMP grade products that have been manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Fiber Drum
Quantity 25 kg
Melting Range 102-106°C
CAS Max % 100%
Percent Purity 99%