Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BIS TRIS, OmniPur™ Calbiochem™,

Click to view available options
Quantity:
100 g
5 kg
6976-37-0
C8H19NO5
209.242
OWMVSZAMULFTJU-UHFFFAOYSA-N
81462
CHEBI:41250
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C(CO)N(CCO)C(CO)(CO)CO
6976-37-0
C8H19NO5
209.242
OWMVSZAMULFTJU-UHFFFAOYSA-N
81462
CHEBI:41250
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C(CO)N(CCO)C(CO)(CO)CO
Melting Point 102°C to 106°C
Assay Percent Range ≥99.0%
Solubility Information Soluble in water (1.0%)
Grade Molecular Biology
Quantity 100 g
DNase None Detected
Protease None Detected
Chemical Name or Material BIS TRIS

Recommended Storage : 15°C to 30°C (Do not freeze).

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit