Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BIS TRIS, OmniPur™ Calbiochem™,

Click to view available options
Quantity:
100 g
5 kg
6976-37-0
C8H19NO5
209.242
OWMVSZAMULFTJU-UHFFFAOYSA-N
81462
CHEBI:41250
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C(CO)N(CCO)C(CO)(CO)CO
6976-37-0
C8H19NO5
209.242
OWMVSZAMULFTJU-UHFFFAOYSA-N
81462
CHEBI:41250
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C(CO)N(CCO)C(CO)(CO)CO
Melting Point 102°C to 106°C
Assay Percent Range ≥99.0%
Solubility Information Soluble in water (1.0%)
Grade Molecular Biology
Quantity 5 kg
DNase None Detected
Protease None Detected
Chemical Name or Material BIS TRIS

Recommended Storage : 15°C to 30°C (Do not freeze).