Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismarck Brown Y, Spectrum™

Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
10114-58-6
10114-58-6
Packaging Amber Glass Bottle
Quantity 25 g
CAS Max % 1
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.