Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Bismuth
SCI_ED

Catalog Number p-4416750
Click to view available options
Quantity:
25 g
100 g
Packaging:
Solid
This item is not returnable. View return policy
7440-69-9
Bi
208.98
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
5359367
CHEBI:33301
bismuth
[Bi]
This item is not returnable. View return policy
7440-69-9
Bi
208.98
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
5359367
CHEBI:33301
bismuth
[Bi]

This product is for educational use - no C of A is available.

Color Red or Silver-White
Density 9.8g/mL
Assay Percent Range >99%
Linear Formula Bi
Quantity 25 g
Formula Weight 208.98
Packaging Solid
Form Solid
Chemical Name or Material Bismuth
ShelfLife 5 years
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit