Alfa Aesar™ Bismuth 2-ethylhexanoate, 99.99+%, 70% w/v in xylene

Manufacturer: Alfa Aesar™ 4152114

Quantity 25g

 View more versions of this product

Catalog No. AA4152114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67874-71-9
    Formula Weight 638.61
    MDL Number MFCD00043803
    UN Number UN1307
    Synonym 2-Ethylhexanoic acid bismuth salt
    Chemical Name or Material Bismuth 2-ethylhexanoate

    Specifications

    Formula Weight 638.61
    Health Hazard 3 GHS P Statement
    P280h-P262
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.
    Quantity 25g
    Physical Form Liquid
    Synonym 2-Ethylhexanoic acid bismuth salt
    Health Hazard 1 Warning
    Concentration 70% w/v in xylene
    MDL Number MFCD00043803
    CAS 67874-71-9
    UN Number UN1307
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H226-H361-H312-H332-H315
    Flammable liquid and vapor.
    Suspected of damaging fertility or the unborn child.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES
    Chemical Name or Material Bismuth 2-ethylhexanoate
    Assay ≥99.99%

    GHS H Statement
    H226-H361-H312-H332-H315
    Flammable liquid and vapor.
    Suspected of damaging fertility or the unborn child.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.

    GHS P Statement
    P280h-P262
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit