Alfa Aesar™ Bismuth aluminum oxide hydrate, 95.5%

Manufacturer: Alfa Aesar™ 40393A1

Quantity 1kg

 View more versions of this product

Catalog No. AA40393A1

  Check Availability
Add to Cart

    Chemical Identifiers

    Formula Weight 771.84 (Anhydrous)
    MDL Number MFCD00798526
    Synonym Aluminum bismuth oxide hydrate; Bismuth aluminate hydrate
    Chemical Name or Material Bismuth aluminum oxide hydrate

    Specifications

    Formula Weight 771.84 (Anhydrous)
    Solubility Information Insoluble in water; Soluble in nitric acid; Typical tap density 0;69g/cm3
    Particle Size typically 2 Micron max
    Physical Form Amorphous Powder
    MDL Number MFCD00798526
    Synonym Aluminum bismuth oxide hydrate; Bismuth aluminate hydrate
    Quantity 1kg
    Chemical Name or Material Bismuth aluminum oxide hydrate
    Assay 95.5%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit