Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Citrate, Powder, USP, 49-54%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
813-93-4
C6H5BiO7
398.08
ANERHPOLUMFRDC-UHFFFAOYSA-K
bismuth(3+) 2-hydroxypropane-1,2,3-tricarboxylate
[Bi+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
813-93-4
C6H5BiO7
398.08
ANERHPOLUMFRDC-UHFFFAOYSA-K
bismuth(3+) 2-hydroxypropane-1,2,3-tricarboxylate
[Bi+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

B1973, 813-93-4, C6H5BiO7

Spectrum™ Bismuth Citrate, Powder, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 10 μg/g
Percent Purity 49% to 54%
Copper (Cu) 10 μg/g
Lead (Pb) 10 μg/g
Packaging Fiber Drum
Silver (Ag) 10 μg/g
Quantity 25 kg