Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth ICP Standard, 1mL = 10mg Bi (10,000ppm Bi), Ricca Chemical

Click to view available options
Quantity:
50mL
100mL
500mL
Packaging:
Poly natural
7440-69-9,7697-37-2,7732-18-5
Bi
208.98
JCXGWMGPZLAOME-UHFFFAOYSA-N
5359367
CHEBI:33301
bismuth
[Bi]
7440-69-9,7697-37-2,7732-18-5
Bi
208.98
JCXGWMGPZLAOME-UHFFFAOYSA-N
5359367
CHEBI:33301
bismuth
[Bi]
CAS Min % 86.0,11.57,0.86
CAS Max % 89.0,12.05,0.89
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 10,000ppm Bi
Equivalence 1 mL = 10 mg Bi
Color Colorless
Quantity 100mL
Chemical Name or Material Bismuth ICP Standard