Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bismuth(III) chloride, 97+%, anhydrous, ACROS Organics™
Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
Specifications
Assay Percent Range | 98% min. (Argentometry) |
Linear Formula | BiCl3 |
Fieser | 07,24 |
Merck Index | 15, 1264 |
Solubility Information | Solubility in water: decomposes. Other solubilities: soluble in acids,soluble in abs.alcohol,acetone and ethyl acetate |
Formula Weight | 315.33 |
Physical Form | Crystalline Powder and/or Chunks |
Percent Purity | 97+% |
Density | 4.7500g/mL |
Packaging | Glass bottle |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 232-123-2