Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bismuth(III) chloride oxide, 99.999% (metals basis)

Click to view available options
Quantity:
10g
50g
7787-59-9
BiClHO
261.437
MFCD00010895
BWOROQSFKKODDR-UHFFFAOYSA-N
bismuth oxychloride,mearlite,pearl white,bismuthine, chlorooxo,wismutchloridoxid,bismut-chlorid-oxid,biju,bismuth chloride oxide,basic bismuth chloride,wismut iii-chlorid-oxid
6328152
oxobismuth;hydrochloride
O=[Bi].Cl
7787-59-9
BiClHO
261.437
MFCD00010895
BWOROQSFKKODDR-UHFFFAOYSA-N
bismuth oxychloride,mearlite,pearl white,bismuthine, chlorooxo,wismutchloridoxid,bismut-chlorid-oxid,biju,bismuth chloride oxide,basic bismuth chloride,wismut iii-chlorid-oxid
6328152
oxobismuth;hydrochloride
O=[Bi].Cl
Linear Formula BiOCl
Merck Index 14,1263
Formula Weight 260.43
Physical Form Powder
Percent Purity 99.999%
Density 7.72 g/mL
Odor Odorless
Refractive Index 2.15
Color Brown
Melting Point (decomposition)
Quantity 10g
Chemical Name or Material Bismuth(III) chloride oxide
Assay (metals basis)
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 232-122-7

RTECSNumber : EB2700000

TSCA : Yes

Recommended Storage : Ambient temperatures