Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Bismuth(III) chloride, ultra dry, 99.997+% (metals basis)
Click to view available options
Quantity:
5g
25g
Description
- Ampouled under argon
Specifications
UN Number | UN3260 |
Merck Index | 14,1262 |
Solubility Information | It is soluble in methanol, diethyl ether, acetone. |
Formula Weight | 315.34 |
Physical Form | Powder |
Percent Purity | ≥99.997% |
Density | 4.75 g/mL |
Packaging | Ampule |
Sensitivity | Hygroscopic |
Boiling Point | ~430°C (sublimation) |
Show More |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 232-123-2
RTECSNumber : EB2690000
TSCA : Yes
storageNote1 :
- Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen