Learn More
Bismuth(III) chloride, ultra dry, 99.997+% (metals basis), Thermo Scientific Chemicals
Description
Bismuth chloride is used as a catalyst in organic synthesis. In particular, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. The addition of other metal iodides increases its catalytic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Boiling Point | ∼430°C (sublimation) |
Melting Point | 230°C to 232°C |
Color | White |
Physical Form | Powder |
Quantity | 5 g |
UN Number | UN3260 |
Merck Index | 14,1262 |
Solubility Information | It is soluble in methanol, diethyl ether, acetone. |
Formula Weight | 315.34 |
Percent Purity | ≥99.997% |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
EINECSNumber | 232-123-2 |
RTECSNumber | EB2690000 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.