Bismuth(III) chloride, White powder, 99.99% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1711522

Quantity 100g

 View more versions of this product

Catalog No. AA1711522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 232-123-2
    Physical Form Powder
    Assay Percent Range 99.99% (Metals basis)
    Molecular Formula BiCl3
    Formula Weight 315.34
    MDL Number MFCD00003461
    Chemical Name or Material Bismuth(III) chloride

    Specifications

    Physical Form Powder
    Formula Weight 315.34
    Quantity 100g
    Color White
    DOT Information DOT Class: 8, Packaging Group: III
    EINECS Number 232-123-2
    Odor Irritating
    Assay Percent Range 99.99% (Metals basis)
    Molecular Formula BiCl3
    MDL Number MFCD00003461
    CAS 232-123-2
    Health Hazard 1 H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Bismuth(III) chloride

    H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit