Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bismuth(III) fluoride, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1184422

Quantity 100g

 View more versions of this product

Catalog No. AA1184422

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7787-61-3
MFCD00010893
14,1265
265.98
UN3260
Bismuth(III) fluoride

Specifications

7787-61-3
265.98
UN3260
−200 Mesh
1.74
727°C
Bismuth(III) fluoride
Powder
MFCD00010893
14,1265
Odorless
Hygroscopic
100g
99.999%, (metals basis)

Safety and Handling

GHS H Statement
H314-H319

GHS P Statement
P260-P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P304+P340-P310-P363-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit