Alfa Aesar™ Bismuth(III) isopropoxide

Manufacturer: Alfa Aesar™ 4393806

Quantity 5g

 View more versions of this product

Catalog No. AA4393806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15049-67-9
    Molecular Formula C9H21BiO3
    Linear Formula Bi[OCH(CH3)2]3
    Formula Weight 386.24
    MDL Number MFCD00799065
    UN Number UN3181
    Chemical Name or Material Bismuth (III) isopropoxide

    Specifications

    Formula Weight 386.24
    Health Hazard 3 GHS P Statement
    P210-P280g-P305+P351+P338-P337+P313
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Health Hazard 1 Warning
    Storage Note 1
    • Packaged under Argon
    Quantity 5g
    Physical Form Powder
    Molecular Formula C9H21BiO3
    Linear Formula Bi[OCH(CH3)2]3
    MDL Number MFCD00799065
    Color Dark gray
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    Sensitivity Moisture Sensitive
    CAS 15049-67-9
    Health Hazard 2 GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    UN Number UN3181
    Chemical Name or Material Bismuth (III) isopropoxide

    GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P280g-P305+P351+P338-P337+P313
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    • Packaged under Argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit