Learn More
Bismuth(III) nitrate pentahydrate, ACS, 98% min, Thermo Scientific Chemicals
Description
Luminous paints, preparation of bismuth saltsBismuth(III) nitrate pentahydrate is an oxidant for a variety of 4-substituted Hantzsch 1,4-dihydropyridines. It also serves as a reagent for selective oxidation of sufides to sulfoxides. Further, it is involved in the preparation of Dragendorff reagent and used as a TLC stain. It is also used to prepare other bismuth compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 30°C (decomposition) |
Physical Form | Crystalline |
Quantity | 250 g |
Assay Percent Range | 98.0% |
Solubility Information | Soluble in dilute HNO3 acid solution. Decomposes in water to subnitrate. Soluble in dilute acetic acid,glycerol,acetone. Insoluble in alcohol,ethyl acetateSoluble in acetone,acetic acid and glycerol. Insoluble in ethanol and ethyl acetate. |
IUPAC Name | bismuth(3+) pentahydrate trinitrate |
Formula Weight | 485.07 (394.99 Anhydrous) |
Grade | ACS |
Sensitivity | Hygroscopic |
Appearance | Crystalline |
Show More |
Safety and Handling
Hazard Category | H272-H315-H319-H335-H373 |
Precautionary Statement | P201-P202-P210-P220-P221-P261-P262-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c |
DOTInformation | DOT Class: 5.1, Packaging Group: II |
EINECSNumber | 233-791-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.