Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) oxide, 99.9%, extra pure, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
1304-76-3
Bi2O3
465.95
Bismuth trioxide
1304-76-3
Bi2O3
465.95
Bismuth trioxide
Assay Percent Range 99.9%
Linear Formula Bi2O3
Fieser 01,59
Merck Index 15,1273
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids
Formula Weight 465.95
Physical Form Powder
Percent Purity 99.9%
Grade Extra Pure
Chlorine (Cl) 10ppm max.
Density 8.9000g/mL
Loss on Drying 0.2% max.
Packaging Plastic bottle
Sodium (Na) 50ppm max.
Solubility Solubility in water: insoluble. Other solubilities: soluble in acids
Specific Gravity 8.9
Sulfate 10ppm max.
Boiling Point 1890°C
Color Yellow
Melting Point 825.0°C
Quantity 100g
Chemical Name or Material Bismuth(III) oxide
Show More Show Less

EINECSNumber : 215-134-7