Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bismuth(III) oxide, NanoArc™ BI-7300, 99.5+%

Manufacturer:  Alfa Aesar™ 4558222

Quantity 100g

 View more versions of this product

Catalog No. AA4558222


  Check Availability
Add to cart

Specifications

Description

  • X-ray opaque particle
  • NanoArc™ bismuth oxide can be compounded into polymers or coating systems to increase opacity to x-rays,while the small particle size ensures minimal disruption of physical properties
Description & Specifications

Chemical Identifiers

1304-76-3
465.96
14,1273
Bi2O3
MFCD00003462
Bismuth (III) oxide NanoArc™ BI-7300

Specifications

1304-76-3
Bi2O3
MFCD00003462
8.9g/mL
38nm APS
≈1890°C
817°C
Bismuth (III) oxide NanoArc™ BI-7300
Powder
465.96
14,1273
Odorless
18m2/g
Bright yellow
100g
≥99.5%

Safety and Handling

SDS & Safety Documents

RTECSNumber : EB2984460

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit