Alfa Aesar™ Bismuth(III) phosphate, 99%

Manufacturer: Alfa Aesar™ 3938018

Quantity 50g

 View more versions of this product

Catalog No. AA3938018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10049-01-1
    Formula Weight 303.95
    MDL Number MFCD00049721
    Merck Index 14,1275
    Chemical Name or Material Bismuth(III) phosphate

    Specifications

    Formula Weight 303.95
    Odor Odorless
    CAS 10049-01-1
    Merck Index 14,1275
    Quantity 50g
    Physical Form Powder
    MDL Number MFCD00049721
    Solubility Information Insoluble in water and alkali; Soluble in hydrochloric acid
    TSCA TSCA
    Density 6.32g/mL
    Chemical Name or Material Bismuth(III) phosphate
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit