Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Bismuth(III) tert-pentyloxide, 97+%
Click to view available options
Quantity:
1g
5g
Specifications
Quantity | 5g |
Color | Gray |
Melting Point | 180°C (decomposition) |
Boiling Point | 63°C (at 10-4mmHg)(sublimation) |
Linear Formula | Bi(OC5H11)3 |
UN Number | UN3181 |
Formula Weight | 470.41 |
Physical Form | Powder |
Percent Purity | ≥97% |
Chemical Name or Material | Bismuth(III) tert-pentyloxide |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures