Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bismuth Lead Indium Tin Cadmium eutectic ingot, 99.9% (metals basis)

Low melting application: fuses, etc

Manufacturer:  Alfa Aesar™ 40950P2

Quantity 1 lb.

 View more versions of this product

Catalog No. AA40950P2


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD00801081
Bismuth Lead Indium Tin Cadmium eutectic ingot

Specifications

Bi:Pb:In:Sn:Cd; 44.7:22.6:19.1:8.3:5.3 wt%
Ingot
47.2°C
99.9%, (metals basis)
1 lb.
MFCD00801081
Bismuth Lead Indium Tin Cadmium eutectic ingot

Safety and Handling

SDS & Safety Documents

GHS H Statement
H350-H360-H373-H302-H312-H332
May cause cancer.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit