Alfa Aesar™ Bismuth Lead Tin Cadmium eutectic ingot, 99.9% (metals basis)

Low melting application: fuses, etc

Manufacturer: Alfa Aesar™ 40951P2

Quantity 1 lb.

 View more versions of this product

Catalog No. AA40951P2

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00801077
    Chemical Name or Material Bismuth Lead Tin Cadmium eutectic ingot

    Specifications

    Health Hazard 3 GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H350-H360-H373-H302-H332
    May cause cancer.
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    TSCA TSCA
    Physical Form Ingot
    Health Hazard 1 Danger
    Melting Point 70°C
    Avg. Mol. Wt. or Mol. Wt. Range Bi:Pb:Sn:Cd; 50.0:26.7:13.3:10.0 wt%
    Quantity 1 lb.
    MDL Number MFCD00801077
    Chemical Name or Material Bismuth Lead Tin Cadmium eutectic ingot
    Assay 99.9%, (metals basis)

    GHS H Statement
    H350-H360-H373-H302-H332
    May cause cancer.
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit