Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bismuth Lead Tin ingot (Rose's metal)

Manufacturer:  Alfa Aesar™ 4051622

Quantity 100g

 View more versions of this product

Catalog No. AA4051622


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD00804027
Bismuth Lead Tin ingot (Rose's metal)

Specifications

Bi:Pb:Sn; 50:25:25 wt%
Low Melting Alloy
95° to 96°C
(Rose's metal)
100g
MFCD00804027
Bismuth Lead Tin ingot (Rose's metal)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit