Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Needles, 99.99% (Metals basis), Alfa Aesar™

Form: Approx. 1.5-3.0mm diameter; Various lengths

Manufacturer:  Alfa Aesar 41485A3

Quantity 2kg

 View more versions of this product

Catalog No. AA41485A3


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-69-9
99.99% (Metals basis)
Bismuth
Needles
MFCD00134033

Specifications

2kg
Gray needles
Needles
MFCD00134033
Gray
7440-69-9
9.79g/cm3
99.99% (Metals basis)
Odorless
Bismuth
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit