Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth oxychloride, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
7787-59-9
BiClHO
261.437
MFCD00010895
BWOROQSFKKODDR-UHFFFAOYSA-N
bismuth oxychloride,mearlite,pearl white,bismuthine, chlorooxo,wismutchloridoxid,bismut-chlorid-oxid,biju,bismuth chloride oxide,basic bismuth chloride,wismut iii-chlorid-oxid
6328152
oxobismuth;hydrochloride
O=[Bi].Cl
7787-59-9
BiClHO
261.437
MFCD00010895
BWOROQSFKKODDR-UHFFFAOYSA-N
bismuth oxychloride,mearlite,pearl white,bismuthine, chlorooxo,wismutchloridoxid,bismut-chlorid-oxid,biju,bismuth chloride oxide,basic bismuth chloride,wismut iii-chlorid-oxid
6328152
oxobismuth;hydrochloride
O=[Bi].Cl
For ultra trace analysis
Assay Percent Range 99.99%
Linear Formula O=BiCl
Merck Index 15, 1274
Bismuth (Bi) 79.5 to 80.5 %
Formula Weight 260.43
Physical Form Powder
Grade Trace Metal Basis
Packaging Glass bottle
Total Trace Metal Impurities 100ppm max.
Color Gray to White
Quantity 50g
Chemical Name or Material Bismuth(III) oxychloride
Show More Show Less

GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.

GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protectio

Warning