Alfa Aesar™ Bismuth, plasma standard solution, Specpure™, Bi 10,000µg/mL

Manufacturer: Alfa Aesar™ 14388AE

Quantity 100mL

 View more versions of this product

Catalog No. AA14388AE

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00134033
    UN Number UN3264
    Chemical Name or Material Bismuth plasma standard solution, Bi 10000μg/mL
    Grade Specpure™

    Specifications

    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314
    Causes severe skin burns and eye damage.
    Quantity 100mL
    TSCA TSCA
    Physical Form Liquid
    Concentration Bi in 5% HNO3
    MDL Number MFCD00134033
    UN Number UN3264
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
    Chemical Name or Material Bismuth plasma standard solution, Bi 10000μg/mL
    Grade Specpure™

    GHS H Statement
    H314
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit