Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bismuth rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis)

Click to view available options
Quantity:
250g
1kg
7440-69-9
Bi
208.98
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth, elemental,unii-u015tt5i8h,powder,iii ion,shot, elongated,rod, 12.7mm 0.5in dia,atom,needles,pieces,rod
5359367
CHEBI:33301
bismuth
[Bi]
7440-69-9
Bi
208.98
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth, elemental,unii-u015tt5i8h,powder,iii ion,shot, elongated,rod, 12.7mm 0.5in dia,atom,needles,pieces,rod
5359367
CHEBI:33301
bismuth
[Bi]
Quantity 250g
Weight ≈12.5g/cm
Odor Odorless
Physical Form Rod, ≈12.5g/cm
Percent Purity 99.9%
Chemical Name or Material Bismuth rod, 12.7mm (0.5 in.) dia.
Assay (metals basis)

EINECSNumber : 231-177-4

TSCA : Yes

Recommended Storage : Ambient temperatures