Alfa Aesar™ Bismuth rod, Puratronic™, 99.9999% (metals basis)

Manufacturer: Alfa Aesar™ 47229KF

Quantity 1 piece

 View more versions of this product

Catalog No. AA47229KF

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7440-69-9
    MDL Number MFCD00134033
    Chemical Name or Material Bismuth rod, 10mm (0.39 in.) dia. x 145mm (5.71 in.) long
    Grade Puratronic™

    Specifications

    TSCA TSCA
    MDL Number MFCD00134033
    Odor Odorless
    Weight ≈7.5g/cm
    Quantity 1 piece
    CAS 7440-69-9
    Chemical Name or Material Bismuth rod, 10mm (0.39 in.) dia. x 145mm (5.71 in.) long
    Assay 99.9999%, (metals basis)
    Grade Puratronic™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit