Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Subcarbonate, USP, 97.6-100.7%, Spectrum™

Click to view available options
Quantity:
25 g
50 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5892-10-4
CBi2O5
509.97
MGLUJXPJRXTKJM-UHFFFAOYSA-L
({[(oxobismuthanyl)oxy]carbonyl}oxy)bismuthanone
O=[Bi]OC(=O)O[Bi]=O
5892-10-4
CBi2O5
509.97
MGLUJXPJRXTKJM-UHFFFAOYSA-L
({[(oxobismuthanyl)oxy]carbonyl}oxy)bismuthanone
O=[Bi]OC(=O)O[Bi]=O

B1969, 5892-10-4, Approx. (BiO)2CO3

Spectrum™ Bismuth Subcarbonate, USP is used as a filler in radiopaque catheters which can be seen by x-ray. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 5 ppm
Percent Purity 97.6 to 100.7%
Chloride 0.0005
Copper (Cu) 5e-005
Lead (Pb) 2e-005
Loss on Drying 0.01
Nitrate 0.004
Packaging Fiber Drum
Silver (Ag) 2.5e-005
Quantity 45 kg
Show More Show Less