Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Subgallate, USP, 52-57%, Spectrum™ Chemical
SDP

Catalog Number 18601010
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
99-26-3
C7H5BiO6
394.09
MFCD00044980
JAONZGLTYYUPCT-UHFFFAOYSA-K
2,7-dihydroxy-2H-benzo[d]1,3-dioxa-2-bismacyclopentane-5-carboxylic acid
O[Bi]1OC2=C(O1)C(O)=CC(=C2)C(O)=O
99-26-3
C7H5BiO6
394.09
MFCD00044980
JAONZGLTYYUPCT-UHFFFAOYSA-K
2,7-dihydroxy-2H-benzo[d]1,3-dioxa-2-bismacyclopentane-5-carboxylic acid
O[Bi]1OC2=C(O1)C(O)=CC(=C2)C(O)=O

B1116, 99-26-3, C7H5BiO6

Spectrum™ Chemical Bismuth Subgallate, USP is used to treat malodor by deodorizing flatulence and stool odors. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 500 g
CAS Max % 100%
Grade USP
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit