Learn More
Description
Bismuth Subgallate USP is an amorphous, bright yellow powder that is odorless. It is stable in air, but affected by light.
Alternative name(s): Gallic acid bismuth basic salt
Specifications
Specifications
| Chemical Name or Material | Bismuth Subgallate USP |
| Packaging | Jar, 100mL, White, HDPE, Round, 38-400, TE Pano Neck |
| CAS | 99-26-3 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 394.091 |
| Quantity | 25 g |
| Percent Purity | 52 to 57 % as Bi2O3 |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
