Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Subnitrate, Powder, USP, 79%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
1304-85-4
BiN3O9
394.99
PPNKDDZCLDMRHS-UHFFFAOYSA-N
bis(nitrooxy)bismuthanyl nitrate
[O-][N+](=O)O[Bi](O[N+]([O-])=O)O[N+]([O-])=O
1304-85-4
BiN3O9
394.99
PPNKDDZCLDMRHS-UHFFFAOYSA-N
bis(nitrooxy)bismuthanyl nitrate
[O-][N+](=O)O[Bi](O[N+]([O-])=O)O[N+]([O-])=O

BI150, 1304-85-4, Bi5O(OH)9(NO3)4

Spectrum™ Bismuth Subnitrate, Powder, USP is used as a dermatological agent and antidiarrheal. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
UN Number UN1477
Arsenic (As) 8 ppm
Percent Purity 79%
Chloride 0.035%
Loss on Drying 3%
Packaging Amber Glass Bottle
Quantity 500 g