Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Subsalicylate, USP, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
14882-18-9
C7H6BiO4
363.10
QBWLKDFBINPHFT-UHFFFAOYSA-L
4-oxo-2H,4H-benzo[d]1,3-dioxa-2-bismacyclohexan-2-yl hydrate
O.O=C1O[Bi]OC2=CC=CC=C12
14882-18-9
C7H6BiO4
363.10
QBWLKDFBINPHFT-UHFFFAOYSA-L
4-oxo-2H,4H-benzo[d]1,3-dioxa-2-bismacyclohexan-2-yl hydrate
O.O=C1O[Bi]OC2=CC=CC=C12

B1703, 14882-18-9, C7H5BiO4

Spectrum™ Bismuth Subsalicylate, USP is an antacid medication used to treat temporary discomforts of the stomach and gastrointestinal tract. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 10 μg/g
Bismuth (Bi) 56.0-59.4%
Copper (Cu) 10 μg/g
Lead (Pb) 10 μg/g
Loss on Drying 0.01
Nitrate 0.004
Packaging Poly Pail
pH Range 2.7-5.0
Silver (Ag) 10 μg/g
Quantity 500 g
Show More Show Less