Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Subsalicylate, USP, Spectrum™ Chemical
SDP

Catalog Number 18601023
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
14882-18-9
C7H6BiO4
363.10
QBWLKDFBINPHFT-UHFFFAOYSA-L
4-oxo-2H,4H-benzo[d]1,3-dioxa-2-bismacyclohexan-2-yl hydrate
O.O=C1O[Bi]OC2=CC=CC=C12
14882-18-9
C7H6BiO4
363.10
QBWLKDFBINPHFT-UHFFFAOYSA-L
4-oxo-2H,4H-benzo[d]1,3-dioxa-2-bismacyclohexan-2-yl hydrate
O.O=C1O[Bi]OC2=CC=CC=C12

B1703, 14882-18-9, C7H5BiO4

Spectrum™ Chemical Bismuth Subsalicylate, USP is an antacid medication used to treat temporary discomforts of the stomach and gastrointestinal tract. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
pH 2.7-5.0
CAS Max % 100%
Quantity 12 kg
Grade USP
Loss on Drying 1%
Packaging Fiber Drum
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit